This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SDF2 SIRNA Assays ›
Gene Symbol:
Sdf2 Supt6
Gene Name:
stromal cell derived factor 2
suppressor of Ty 6
Gene Aliases:
AI853825
5131400N11Rik, AI132449, SPT6, Supt6h, mKIAA0162
Chromosome Location:
Chr.11: 78245746 - 78255496 on Build GRCm38
Chromosome Location:
Chr.11: 78206749 - 78245703 on Build GRCm38
UniGene ID:
Mm.20755
Mm.20755
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID MSS208859
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sdf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009143.3 NP_033169.3 1 391
GenBank mRNA AK002496.1 - 1 147
BC058798.1 - 1 391 6485217
BC062881.1 - 1 137 5693309
D50646.1 - 1 145
DN176017.1 - 1 391
Gene Symbol: Supt6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK136094.1 - 204

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0