This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAD50 SIRNA Assays ›
Gene Symbol:
Rad50
Gene Name:
RAD50 homolog (S. cerevisiae)
Gene Aliases:
Mrell, Rad50l
Chromosome Location:
Chr.11: 53649519 - 53707319 on Build GRCm38
UniGene ID:
Mm.4888
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS208387
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rad50
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009012.2 NP_033038.2 9, 10 1711
GenBank mRNA AK037641.1 - 9, 10 1712
AK087982.1 - 1, 2 122
U66887.1 - 9, 10 1689

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of mitotic recombination
telomere maintenance
DNA repair
double-strand break repair
DNA recombination
cellular response to DNA damage stimulus
telomere maintenance via telomerase
cell cycle
regulation of mitotic cell cycle
positive regulation of protein autophosphorylation
DNA duplex unwinding
positive regulation of kinase activity
negative regulation of viral entry into host cell
chromosome organization
protein heterooligomerization
meiotic cell cycle
chromosome organization involved in meiotic cell cycle
nucleic acid phosphodiester bond hydrolysis
single-stranded DNA endodeoxyribonuclease activity
nucleotide binding
DNA binding
double-stranded DNA binding
ATP-dependent DNA helicase activity
ATP binding
3'-5' exonuclease activity
hydrolase activity
ATPase activity
protein binding, bridging
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0