This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDRG1 SIRNA Assays ›
Gene Symbol:
Ndrg1
Gene Name:
N-myc downstream regulated gene 1
Gene Aliases:
CAP43, CMT4D, DRG1, HMSNL, NMSL, Ndr1, Ndrl, PROXY1, RTP, TDD5
Chromosome Location:
Chr.15: 66929318 - 66969641 on Build GRCm38
UniGene ID:
Mm.30837
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS206976
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ndrg1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008681.2 NP_032707.2 8 638
GenBank mRNA AK004548.1 - 8 638
AK098075.1 - 8 637
AK161860.1 - 8 640
AK165007.1 - 8 638
AK170281.1 - 8 639
AK170439.1 - 8 637
AK170636.1 - 8 637
AK170809.1 - 8 638
BC015282.1 - 8 593 4237943
BC071235.1 - 8 646 4242075
BC141246.1 - 8 529 9055858
BC171968.1 - 8 529 9054254
CA320478.1 - 5 381
U52073.1 - 8 614
U60593.1 - 8 636

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
peripheral nervous system myelin maintenance
mast cell activation
positive regulation of spindle checkpoint
microtubule binding
Rab GTPase binding
gamma-tubulin binding
cadherin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0