This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NCK1 SIRNA Assays ›
Gene Symbol:
Nck1
Gene Name:
non-catalytic region of tyrosine kinase adaptor protein 1
Gene Aliases:
6330586M15Rik, D230010O13Rik, Nck, Nck-1
Chromosome Location:
Chr.9: 100494981 - 100546053 on Build GRCm38
UniGene ID:
Mm.181485
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS206957
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nck1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010878.2 NP_035008.2 3 447
GenBank mRNA AF043259.1 - 3 535
AF084183.1 - 2 370
AK053155.1 - 244
AK054051.1 - 2 276
AK210839.1 - 1 64
BC002015.1 - 3 447 3593441

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of translation
negative regulation of protein kinase activity
substrate-dependent cell migration, cell extension
actin filament organization
cell migration
lamellipodium assembly
positive regulation of actin filament polymerization
negative regulation of peptidyl-serine phosphorylation
positive regulation of translation in response to endoplasmic reticulum stress
positive regulation of T cell proliferation
T cell activation
positive regulation of transcription from RNA polymerase II promoter
response to other organism
negative regulation of cell death
peptidyl-serine dephosphorylation
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of cap-dependent translational initiation
positive regulation of cap-independent translational initiation
negative regulation of PERK-mediated unfolded protein response
negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
protein kinase inhibitor activity
receptor binding
protein binding
protein kinase regulator activity
protein domain specific binding
receptor tyrosine kinase binding
eukaryotic initiation factor eIF2 binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0