This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANAPC1 SIRNA Assays ›
Gene Symbol:
Anapc1
Gene Name:
anaphase promoting complex subunit 1
Gene Aliases:
2610021O03Rik, AI047775, AI853536, AW547281, Apc1, Mcpr, tsg24
Chromosome Location:
Chr.2: 128610083 - 128687410 on Build GRCm38
UniGene ID:
Mm.277408
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS206578
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Anapc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008569.2 NP_032595.2 40 5098
GenBank mRNA AK034858.1 - 8 629
AK090134.1 - 40 5080
BC139002.1 - 40 5079 8862024
BC145492.1 - 40 5079 9054055
X80169.1 - 40 5083

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
cell division
protein K11-linked ubiquitination
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0