This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MATR3 SIRNA Assays ›
Gene Symbol:
Matr3
Gene Name:
matrin 3
Gene Aliases:
1110061A14Rik, 2810017I02Rik, AI841759, AW555618, D030046F20Rik, mKIAA0723
Chromosome Location:
Chr.18: 35562158 - 35592045 on Build GRCm38
UniGene ID:
Mm.215034
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS206532
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Matr3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010771.6 NP_034901.2 8 1724
GenBank mRNA AK029510.1 - 7 1102
AK087939.1 - 8 1609
AK088351.1 - 8 1617
AK129199.1 - 8 1675
AK135937.1 - 7 1457
AK148384.1 - 1 51
AK157744.1 - 7 820
AK161128.1 - 8 1623
AK182890.1 - 1 114
BC029070.1 - 8 1635 3989914
BC090833.1 - 7 1497 6854095
CT010322.1 - 7 1408

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_199189, NM_018834, NM_001194956, NM_001282278, NM_001194954, and others.
This siRNA also targets the following transcripts in Rat: NM_019149, XM_006254534, XM_006254535, AB205483, M63485, and others.

Gene Ontology Categories:

Function(s) Process(es)

posttranscriptional regulation of gene expression
nucleotide binding
nucleic acid binding
RNA binding
zinc ion binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header