This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNK5 SIRNA Assays ›
Gene Symbol:
Kcnk5
Gene Name:
potassium channel, subfamily K, member 5
Gene Aliases:
KCNK5b, TASK-2
Chromosome Location:
Chr.14: 20140057 - 20181782 on Build GRCm38
UniGene ID:
Mm.68998
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS205718
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnk5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021542.4 NP_067517.1 1, 2 507
GenBank mRNA AF259395.1 - 1, 2 177
AF319542.1 - 1, 2 246
AK085330.1 - 1, 2 508
AK132172.1 - 1, 2 535
AK172608.1 - 1, 2 508
BC034012.1 - 1, 2 509 4239113
BC051088.1 - 1, 2 476
BC058164.1 - 1, 2 498 5323480

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

stabilization of membrane potential
regulation of ion transmembrane transport
cellular potassium ion transport
potassium ion transmembrane transport
voltage-gated potassium channel activity
potassium ion leak channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0