This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ICA1 SIRNA Assays ›
Gene Symbol:
Ica1
Gene Name:
islet cell autoantigen 1
Gene Aliases:
69kDa, ICA69
Chromosome Location:
Chr.6: 8630527 - 8778537 on Build GRCm38
UniGene ID:
Mm.275683
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS205329
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ica1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252266.1 NP_001239195.1 11 1111
NM_010492.3 NP_034622.3 13 1587
GenBank mRNA AB303567.1 - 12 1098
BC002030.1 - 13 1221 3487814
CD775505.1 - 2 500
CO044997.1 - 2 500
DV658174.1 - 7 497
U26459.1 - 13 1210
U26460.1 - 12 1116
U26461.1 - 11 1052
U34595.1 - 11 1083

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
neurotransmitter transport
regulation of protein homodimerization activity
regulation of neurotransmitter secretion
protein domain specific binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0