This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HOXD4 SIRNA Assays ›
Gene Symbol:
Hoxd4
Gene Name:
homeobox D4
Gene Aliases:
6030436D05Rik, Hox-4.2, Hox-5.1
Chromosome Location:
Chr.2: 74721978 - 74729160 on Build GRCm38
UniGene ID:
Mm.482194
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS205193
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hoxd4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010469.2 NP_034599.2 6 1973
GenBank mRNA AK135479.1 - 2 764
AK136751.1 - 771
BC139206.1 - 2 865 8862228
BC139207.1 - 2 865 8862229
J03770.1 - 2 944

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001105885, XM_008761886, XM_008761883, XM_008761885, XM_008761884, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
anterior/posterior pattern specification
embryonic skeletal system morphogenesis
stem cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0