This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HOXD10 SIRNA Assays ›
Gene Symbol:
Hoxd10
Gene Name:
homeobox D10
Gene Aliases:
AI385591, AI874987, Hox-4.5, Hox-5.3
Chromosome Location:
Chr.2: 74684275 - 74695106 on Build GRCm38
UniGene ID:
Mm.24420
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS205187
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hoxd10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013554.5 NP_038582.2 2 986
GenBank mRNA AK134330.1 - 2 988
BC013463.1 - 2 947 4240579
BC048690.1 - 2 929 6516538

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_002148, AW243295, BC069619, BC074760, X59373.
This siRNA also targets the following transcripts in Rat: NM_001107094.

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
single fertilization
skeletal muscle tissue development
adult locomotory behavior
anterior/posterior pattern specification
proximal/distal pattern formation
regulation of gene expression
spinal cord motor neuron cell fate specification
embryonic limb morphogenesis
forelimb morphogenesis
hindlimb morphogenesis
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system morphogenesis
peripheral nervous system neuron development
neuromuscular process
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0