This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HIPK3 SIRNA Assays ›
Gene Symbol:
Hipk3
Gene Name:
homeodomain interacting protein kinase 3
Gene Aliases:
DYRK6, FIST3, PKY, mir-1902
Chromosome Location:
Chr.2: 104426481 - 104494489 on Build GRCm38
UniGene ID:
Mm.257925
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS205130
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hipk3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145824.1 NP_001139296.1 16 3820
NM_010434.2 NP_034564.2 16 3823
GenBank mRNA AF077660.1 - 16 3577
AF170305.1 - 15 3287
AF305238.1 - 15 3285
AK140130.1 - 16 3926
BC131663.1 - 16 3455
BC131664.1 - 16 3451 40110751

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
mRNA transcription
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
negative regulation of apoptotic process
positive regulation of DNA binding
negative regulation of JUN kinase activity
nucleotide binding
DNA binding
transcription corepressor activity
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header