This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GDNF SIRNA Assays ›
Gene Symbol:
Gdnf
Gene Name:
glial cell line derived neurotrophic factor
Gene Aliases:
AI385739
Chromosome Location:
Chr.15: 7810048 - 7837580 on Build GRCm38
UniGene ID:
Mm.4679
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS204652
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gdnf
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301332.1 NP_001288261.1 3 1602
NM_001301333.1 NP_001288262.1 3 964
NM_001301357.1 NP_001288286.1 2 652
NM_010275.3 NP_034405.1 3 1680
GenBank mRNA BC119031.1 - 3 696 8733780
D49921.1 - 2 587
D88264.1 - 3 882
U37459.1 - 3 717

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metanephros development
ureteric bud development
branching involved in ureteric bud morphogenesis
neural crest cell migration
organ induction
postsynaptic membrane organization
mesenchymal to epithelial transition involved in metanephros morphogenesis
transmembrane receptor protein tyrosine kinase signaling pathway
transforming growth factor beta receptor signaling pathway
nervous system development
peripheral nervous system development
cell proliferation
positive regulation of cell proliferation
response to wounding
regulation of gene expression
postganglionic parasympathetic fiber development
neuron differentiation
peristalsis
neuron projection development
positive regulation of monooxygenase activity
negative regulation of neuron apoptotic process
positive regulation of cell differentiation
positive regulation of transcription from RNA polymerase II promoter
mRNA stabilization
enteric nervous system development
sympathetic nervous system development
regulation of dopamine uptake involved in synaptic transmission
ureteric bud formation
regulation of morphogenesis of a branching structure
cellular response to dexamethasone stimulus
regulation of ureteric bud formation
positive regulation of ureteric bud formation
positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
positive regulation of branching involved in ureteric bud morphogenesis
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
transforming growth factor beta receptor binding
growth factor activity
glial cell-derived neurotrophic factor receptor binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0