This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GABRB1 SIRNA Assays ›
Gene Symbol:
Gabrb1
Gene Name:
gamma-aminobutyric acid (GABA) A receptor, subunit beta 1
Gene Aliases:
AW061132, B230208N19Rik, Gabrb-1
Chromosome Location:
Chr.5: 71658056 - 72149572 on Build GRCm38
UniGene ID:
Mm.38567
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS204522
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gabrb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008069.4 NP_032095.1 4 439
GenBank mRNA BC130258.1 - 4 443 40142758
BC157920.1 - 4 505 9007350

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
chloride transport
signal transduction
response to toxic substance
ion transmembrane transport
cellular response to histamine
anion transmembrane transport
GABA-A receptor activity
ion channel activity
extracellular ligand-gated ion channel activity
anion channel activity
chloride channel activity
ligand-gated ion channel activity
GABA receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0