This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other F8 SIRNA Assays ›
Gene Symbol:
F8
Gene Name:
coagulation factor VIII
Gene Aliases:
Cf-8, Cf8, FVIII
Chromosome Location:
Chr.X: 75172715 - 75383525 on Build GRCm38
UniGene ID:
Mm.1805
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS204148
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: F8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161373.1 NP_001154845.1 13 2912
NM_001161374.1 NP_001154846.1 13 2912
NM_007977.2 NP_032003.2 14 3122
GenBank mRNA BC138347.1 - 13 2793 8861367
BC145245.1 - 13 2793 9053797
L05573.1 - 14 3120

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
acute-phase response
blood coagulation
blood coagulation, intrinsic pathway
hemostasis
platelet activation
oxidation-reduction process
serine-type endopeptidase activity
copper ion binding
oxidoreductase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0