This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ETV6 SIRNA Assays ›
Gene Symbol:
Etv6
Gene Name:
ets variant 6
Gene Aliases:
AW123102, AW557856, Tel
Chromosome Location:
Chr.6: 134034583 - 134270158 on Build GRCm38
UniGene ID:
Mm.269995
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS204015
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Etv6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK035231.1 - 6 1118
AK078773.1 - 7 1311
AK154569.1 - 7 1399
AK155150.1 - 7 1584
AK155752.1 - 6 1134
AK157265.1 - 7 1253
BC052163.1 - 7 1272 6513848

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0