This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DSC3 SIRNA Assays ›
Gene Symbol:
Dsc3
Gene Name:
desmocollin 3
Gene Aliases:
5430426I24Rik
Chromosome Location:
Chr.18: 19960930 - 20002413 on Build GRCm38
UniGene ID:
Mm.89935
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS203696
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dsc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291809.1 NP_001278738.1 2 287
NM_007882.3 NP_031908.3 2 287
GenBank mRNA BC144845.1 - 2 272
BY151533.1 - 2 259
BY745760.1 - 3 333
Y11169.1 - 2 287

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
protein stabilization
calcium ion binding
gamma-catenin binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0