This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DSC2 SIRNA Assays ›
Gene Symbol:
Dsc2
Gene Name:
desmocollin 2
Gene Aliases:
AW228162, Dsc2a, Dsc2b
Chromosome Location:
Chr.18: 20030633 - 20059552 on Build GRCm38
UniGene ID:
Mm.280547
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS203694
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dsc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013505.3 NP_038533.1 13 2255
GenBank mRNA AK028604.1 - 14 2295
AK028636.1 - 13 2256
AK030333.1 - 13 2256
AK048547.1 - 13 2256
AK172168.1 - 13 2090
BC004663.1 - 7 1142
BC057867.1 - 13 2035 5327748
L33779.1 - 13 2054

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
cellular response to starvation
cardiac muscle cell-cardiac muscle cell adhesion
bundle of His cell-Purkinje myocyte adhesion involved in cell communication
regulation of heart rate by cardiac conduction
regulation of ventricular cardiac muscle cell action potential
calcium ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0