This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DLL3 SIRNA Assays ›
Gene Symbol:
Dll3
Gene Name:
delta-like 3 (Drosophila)
Gene Aliases:
pu, pudgy
Chromosome Location:
Chr.7: 28293555 - 28301785 on Build GRCm38
UniGene ID:
Mm.12896
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS203584
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dll3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007866.2 NP_031892.2 8 1748
GenBank mRNA AB013440.1 - 8 1735
AK144282.1 - 8 1749
BC052002.1 - 8 1725 6404029

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
in utero embryonic development
cell fate determination
somitogenesis
Notch signaling pathway
multicellular organism development
compartment pattern specification
nervous system development
tissue development
cell differentiation
negative regulation of Notch signaling pathway
paraxial mesoderm development
negative regulation of astrocyte differentiation
negative regulation of neurogenesis
positive regulation of neurogenesis
Notch binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0