This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DGCR2 SIRNA Assays ›
Gene Symbol:
Dgcr2
Gene Name:
DiGeorge syndrome critical region gene 2
Gene Aliases:
9930034O06Rik, DGS-C, Dgsc, Idd, Lan, Sez12, mKIAA0163
Chromosome Location:
Chr.16: 17840355 - 17894098 on Build GRCm38
UniGene ID:
Mm.254515
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS203549
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dgcr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109750.1 NP_001103220.1 6 1027
NM_010048.3 NP_034178.2 6 1036
GenBank mRNA AK042786.1 - 6 958
AK088131.1 - 6 957
AK129070.1 - 7 886
AK133880.1 - 3 325
AK141594.1 - 6 1002
AK145495.1 - 6 953
AK153026.1 - 6 824
AK161022.1 - 1787
AK161359.1 - 6 752
AK163872.1 - 6 989
AK163903.1 - 6 965
AK190458.1 - 1 87
AK194718.1 - 1 86
AK202091.1 - 1 86
BC060636.1 - 6 977 6849298
BC062978.1 - 6 931 30356000
D78641.1 - 6 961
X95480.1 - 6 902

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
response to drug
cognition
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0