This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CTSB SIRNA Assays ›
Gene Symbol:
Ctsb
Gene Name:
cathepsin B
Gene Aliases:
CB
Chromosome Location:
Chr.14: 63122462 - 63145923 on Build GRCm38
UniGene ID:
Mm.236553
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS203346
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ctsb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007798.3 NP_031824.1 4 417
GenBank mRNA AK002456.1 - 4 366
AK083393.1 - 4 417
AK147192.1 - 4 364
AK149884.1 - 4 366
AK151790.1 - 4 364
AK153047.1 - 4 363
AK159992.1 - 4 402
AK167361.1 - 4 398
AK170959.1 - 4 390
AK190209.1 - 3 215
AK190539.1 - 3 215
AK191055.1 - 3 215
AK191391.1 - 3 215
AK196955.1 - 3 215
AK198213.1 - 3 215
AK199268.1 - 3 216
AK202291.1 - 3 215
AK203639.1 - 3 215
BC006656.1 - 4 408 3500700
M14222.1 - 3 322
S69034.1 - 4 413

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
collagen catabolic process
decidualization
regulation of catalytic activity
proteolysis involved in cellular protein catabolic process
negative regulation of cell death
endopeptidase activity
cysteine-type endopeptidase activity
collagen binding
peptidase activity
cysteine-type peptidase activity
hydrolase activity
kininogen binding
protein complex binding
peptide binding
proteoglycan binding
protein self-association

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0