This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other COL17A1 SIRNA Assays ›
Gene Symbol:
Col17a1
Gene Name:
collagen, type XVII, alpha 1
Gene Aliases:
BP180, Bpag, Bpag2
Chromosome Location:
Chr.19: 47646341 - 47692042 on Build GRCm38
UniGene ID:
Mm.1225
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS203127
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Col17a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290825.1 NP_001277754.1 19 1961
NM_007732.2 NP_031758.2 19 1961
GenBank mRNA BC125031.1 - 19 1961 40086689
BC125032.1 - 19 1960 40086691
CF734205.1 - 6 643
L08407.1 - 19 2013

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0