This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CAPN6 SIRNA Assays ›
Gene Symbol:
Capn6
Gene Name:
calpain 6
Gene Aliases:
-
Chromosome Location:
Chr.X: 143802236 - 143827412 on Build GRCm38
UniGene ID:
Mm.30290
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS202594
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Capn6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007603.3 NP_031629.3 8 1298
GenBank mRNA AK145116.1 - 8 1313
BC052167.1 - 8 1199 6517565
Y12582.1 - 8 1207

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031808, AF067793.

Gene Ontology Categories:

Function(s) Process(es)

microtubule bundle formation
proteolysis
regulation of cytoskeleton organization
calcium-dependent cysteine-type endopeptidase activity
protein binding
microtubule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0