This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BTN1A1 SIRNA Assays ›
Gene Symbol:
Btn1a1
Gene Name:
butyrophilin, subfamily 1, member A1
Gene Aliases:
Btn
Chromosome Location:
Chr.13: 23456984 - 23465914 on Build GRCm38
UniGene ID:
Mm.1420
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS202453
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Btn1a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013483.3 NP_038511.1 8 1582
GenBank mRNA AK142596.1 - 8 1489
AK145168.1 - 8 1583
BC011497.1 - 8 1556 3583827
BC031459.1 - 7 1480 3582574
S80642.1 - 6 1308

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cellular metabolic process
negative regulation of activated T cell proliferation
negative regulation of cytokine secretion
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0