This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BNIP3 SIRNA Assays ›
Gene Symbol:
Bnip3
Gene Name:
BCL2/adenovirus E1B interacting protein 3
Gene Aliases:
Nip3
Chromosome Location:
Chr.7: 138890836 - 138909506 on Build GRCm38
UniGene ID:
Mm.378890
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS202423
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bnip3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009760.4 NP_033890.1 5 533
GenBank mRNA AF041054.1 - 5 503
AK014223.1 - 5 534
AK075943.1 - 5 534
AK152610.1 - 5 529
AK170865.1 - 5 497
BC033278.1 - 5 441
BC046603.1 - 5 523 6390074

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053420, FQ215127, BC070958, FQ215108, KC660101, and others.

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
apoptotic process
cell death
granzyme-mediated apoptotic signaling pathway
negative regulation of mitochondrial fusion
regulation of mitochondrion organization
mitochondrial protein catabolic process
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of programmed cell death
positive regulation of protein complex disassembly
mitochondrial fragmentation involved in apoptotic process
negative regulation of membrane potential
regulation of mitochondrial membrane permeability
autophagic cell death
brown fat cell differentiation
neuron apoptotic process
defense response to virus
cellular response to hydrogen peroxide
cellular response to cobalt ion
cellular response to hypoxia
reactive oxygen species metabolic process
positive regulation of mitochondrial fission
positive regulation of release of cytochrome c from mitochondria
intrinsic apoptotic signaling pathway
mitochondrial outer membrane permeabilization
toxin transport
regulation of aerobic respiration
intrinsic apoptotic signaling pathway in response to hypoxia
negative regulation of reactive oxygen species metabolic process
protein binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0