This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BOC SIRNA Assays ›
Gene Symbol:
Boc
Gene Name:
biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein
Gene Aliases:
4732455C11, mFLJ00376
Chromosome Location:
Chr.16: 44485045 - 44558870 on Build GRCm38
UniGene ID:
Mm.41561
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS202049
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Boc
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172506.2 NP_766094.1 12 2560
GenBank mRNA AF388037.1 - 11 2013
AK028770.1 - 12 2581
AK028889.1 - 12 2561
AK131182.1 - 7 4780
BC056138.1 - 3 463
BC062892.1 - 12 2540 6418202
BC078631.1 - 12 2504 6515418

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
smoothened signaling pathway
axon guidance
regulation of striated muscle tissue development
single organismal cell-cell adhesion
cell projection organization
positive regulation of myoblast differentiation
mitophagy in response to mitochondrial depolarization
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0