This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALPL SIRNA Assays ›
Gene Symbol:
Alpl
Gene Name:
alkaline phosphatase, liver/bone/kidney
Gene Aliases:
APTNAP, Akp-2, Akp2, TNAP, TNSALP
Chromosome Location:
Chr.4: 137741731 - 137796384 on Build GRCm38
UniGene ID:
Mm.288186
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS201884
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Alpl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287172.1 NP_001274101.1 7 1045
NM_001287176.1 NP_001274105.1 7 1168
NM_007431.3 NP_031457.2 7 1020
GenBank mRNA AB473959.1 - 6 784
AK076459.1 - 7 955
AK161780.1 - 7 956
AK164002.1 - 7 1013
AK166732.1 - 7 954
AK167023.1 - 7 955
AK167481.1 - 7 955
BC065175.1 - 7 943 6807509
J02980.1 - 7 959
X13409.1 - 7 903

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
endochondral ossification
developmental process involved in reproduction
metabolic process
dephosphorylation
response to antibiotic
response to glucocorticoid
cellular response to organic cyclic compound
catalytic activity
alkaline phosphatase activity
protein binding
pyrophosphatase activity
hydrolase activity
phosphatase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0