This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDE6C SIRNA Assays ›
Gene Symbol:
Pde6c
Gene Name:
phosphodiesterase 6C, cGMP specific, cone, alpha prime
Gene Aliases:
cpfl1
Chromosome Location:
Chr.19: 38132781 - 38183955 on Build GRCm38
UniGene ID:
Mm.450141
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS201413
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pde6c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170959.1 NP_001164430.1 5, 6 1094
NM_033614.2 NP_291092.1 5, 6 1094
GenBank mRNA AF411063.1 - 5, 6 929
BC027050.1 - 5, 6 1094 5358611

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108522, XM_006231329, XM_006231328.

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
visual perception
phototransduction, visible light
retinal cone cell development
response to stimulus
sensory perception of light stimulus
nucleotide binding
3',5'-cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
cGMP binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0