This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STON2 SIRNA Assays ›
Gene Symbol:
Ston2
Gene Name:
stonin 2
Gene Aliases:
4933401N24Rik, AI425999, AW544599
Chromosome Location:
Chr.12: 91633009 - 91786436 on Build GRCm38
UniGene ID:
Mm.23125
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS201041
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ston2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175367.6 NP_780576.1 7 3143
GenBank mRNA AK036612.1 - 7 3144
BC132531.1 - 6 2729 40130808
BC138054.1 - 6 2729 8861073

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001135874, XM_006240406, XM_006240407.

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
intracellular protein transport
endocytosis
chemical synaptic transmission
vesicle-mediated transport
regulation of endocytosis
synaptic vesicle endocytosis
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0