This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLCO4A1 SIRNA Assays ›
Gene Symbol:
Slco4a1
Gene Name:
solute carrier organic anion transporter family, member 4a1
Gene Aliases:
OATP-E, Slc21a12
Chromosome Location:
Chr.2: 180456250 - 180474853 on Build GRCm38
UniGene ID:
Mm.133687
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS200974
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slco4a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148933.1 NP_683735.1 10 2169
GenBank mRNA AK033598.1 - 10 1989
AK172688.1 - 10 1919
BC026987.1 - 1 41
BC030719.1 - 10 2169 4501966
BC030720.1 - 10 2118 4505508
BC033602.1 - 3 371

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
organic anion transport
thyroid hormone metabolic process
thyroid hormone transport
anion transmembrane transport
transporter activity
organic anion transmembrane transporter activity
thyroid hormone transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0