This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC14A1 SIRNA Assays ›
Gene Symbol:
Slc14a1
Gene Name:
solute carrier family 14 (urea transporter), member 1
Gene Aliases:
2610507K20Rik, 3021401A05Rik, UT-B, Utb1
Chromosome Location:
Chr.18: 78100091 - 78142119 on Build GRCm38
UniGene ID:
Mm.33832
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS200927
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc14a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171010.1 NP_001164481.1 7, 8 1216
NM_001171011.1 NP_001164482.1 6, 7 951
NM_028122.4 NP_082398.1 6, 7 1011
GenBank mRNA AF448798.1 - 5, 6 808
AJ420967.1 - 6, 7 870
AK012066.1 - 6, 7 861
AK041979.1 - 6, 7 953
AK153891.1 - 7, 8 1121
BC058594.1 - 6, 7 898 5364683
BC086673.1 - 6, 7 907 30646772
BC100570.1 - 6, 7 898 30044353

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
water transport
urea transport
transmembrane transport
urea transmembrane transport
water transmembrane transporter activity
urea transmembrane transporter activity
urea channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header