This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WWP1 SIRNA Assays ›
Gene Symbol:
Wwp1
Gene Name:
WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases:
8030445B08Rik, AIP5, SDRP1, Tiul1
Chromosome Location:
Chr.4: 19608296 - 19709004 on Build GRCm38
UniGene ID:
Mm.474845
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS200824
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wwp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276292.1 NP_001263221.1 16 1932
NM_177327.5 NP_796301.2 17 2094
GenBank mRNA AK031762.1 - 1 27
AK032232.1 - 16 1760
AK033138.1 - 9 908
AK137588.1 - 17 2019
AK168567.1 - 17 1958
BC026829.1 - 16 1870 4209397
BC150812.1 - 16 1929 9087723

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
T cell differentiation
lung development
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of transcription, DNA-templated
ubiquitin-protein transferase activity
protein binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0