This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRIM68 SIRNA Assays ›
Gene Symbol:
Trim68
Gene Name:
tripartite motif-containing 68
Gene Aliases:
AI894158, F730114J12Rik, Rnf137, SS-56
Chromosome Location:
Chr.7: 102677579 - 102687329 on Build GRCm38
UniGene ID:
Mm.39043
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS200184
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trim68
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK089546.1 - 2 447
AK155267.1 - 2 447
AK164544.1 - 2 392
BC034249.1 - 2 375 3675490

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008759840.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0