This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLHL21 SIRNA Assays ›
Gene Symbol:
KLHL21
Gene Name:
kelch like family member 21
Gene Aliases:
-
Chromosome Location:
Chr.1: 6590724 - 6602898 on Build GRCh38
UniGene ID:
Hs.7764
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS190705
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KLHL21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324309.1 NP_001311238.1 2 1139
NM_014851.3 NP_055666.2 2 1139
GenBank mRNA AB007938.1 - 2 1242
AK075305.1 - 2 1110
AK090472.1 - 2 908
AK307564.1 - 2 99
BC034039.1 - 2 1110 4811534
BC064980.1 - 2 1092
BC091648.1 - 2 1092 6373903

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic nuclear division
protein ubiquitination
regulation of cytokinesis
chromosome passenger complex localization to spindle midzone
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
cell division
ubiquitin-protein transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0