This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CPNE6 SIRNA Assays ›
Gene Symbol:
CPNE6
Gene Name:
copine 6
Gene Aliases:
-
Chromosome Location:
Chr.14: 24070837 - 24078100 on Build GRCh38
UniGene ID:
Hs.6132
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS190244
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CPNE6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001280558.1 NP_001267487.1 6 754
NM_006032.3 NP_006023.1 5 742
XM_005268216.1 XP_005268273.1 6 698
XM_005268217.1 XP_005268274.1 4 392
GenBank mRNA AB009288.1 - 5 542
AK125195.1 - 4 1304
AK222675.1 - 5 502
AK293655.1 - 6 754
AK294460.1 - 4 657
AK314182.1 - 5 555
AL137688.1 - 3 1953
BC018046.1 - 5 548 4799650

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
chemical synaptic transmission
nervous system development
vesicle-mediated transport
glycerophospholipid biosynthetic process
cellular response to calcium ion
positive regulation of dendrite extension
phosphatidylserine binding
transporter activity
calcium ion binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0