This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C1ORF158 SIRNA Assays ›
Gene Symbol:
C1orf158
Gene Name:
chromosome 1 open reading frame 158
Gene Aliases:
-
Chromosome Location:
Chr.1: 12746186 - 12761169 on Build GRCh38
UniGene ID:
Hs.98095
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS190189
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C1orf158
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152290.3 NP_689503.3 1, 2 362
GenBank mRNA AK026705.1 - 1, 2 329
AK298760.1 - 1, 2 324
BC029894.1 - 1, 2 333 5171302
HY002971.1 - 1, 2 363

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0