This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BCL7B SIRNA Assays ›
Gene Symbol:
BCL7B
Gene Name:
BCL tumor suppressor 7B
Gene Aliases:
-
Chromosome Location:
Chr.7: 73536353 - 73557735 on Build GRCh38
UniGene ID:
Hs.647051
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS190144
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BCL7B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197244.1 NP_001184173.1 4 443
NM_001301061.1 NP_001287990.1 6 736
NM_001707.3 NP_001698.2 5 614
NR_036682.1 5 597
GenBank mRNA AJ223979.1 - 5 493
AK290091.1 - 5 522
AL525884.3 - 6 677
AL553260.3 - 5 880
BC000956.2 - 5 573 3450946
BC001967.2 - 5 480 3461829
BC009548.1 - 5 524 3897805
BI603803.1 - 5 610
BI837138.1 - 4 414
BQ109412.1 - 5 546
X89985.1 - 5 494

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
biological_process
Wnt signaling pathway
cell differentiation
actin binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0