This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDC123 SIRNA Assays ›
Gene Symbol:
CDC123
Gene Name:
cell division cycle 123
Gene Aliases:
C10orf7, D123
Chromosome Location:
Chr.10: 12195962 - 12250590 on Build GRCh38
UniGene ID:
Hs.412842
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS189632
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CDC123
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006023.2 NP_006014.2 11 1081
XM_005252638.3 XP_005252695.1 10 802
GenBank mRNA AK023925.1 - 1 601
AK225634.1 - 11 1047
AK289362.1 - 11 818
AW006077.1 - 3 465
BC001600.1 - 11 831 3542611
BC009598.1 - 11 846 4134376
CR456966.1 - 11 797
D14878.1 - 11 1077
U27112.1 - 11 859

Back To Top

More Information


Gene Ontology Categories:

Function(s)

formation of cytoplasmic translation initiation complex
regulation of translation
cell cycle arrest
regulation of mitotic cell cycle
positive regulation of cell proliferation
positive regulation of translational initiation
cell division
eukaryotic translation initiation factor 2 complex assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0