This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SCEL SIRNA Assays ›
Gene Symbol:
SCEL
Gene Name:
sciellin
Gene Aliases:
-
Chromosome Location:
Chr.13: 77535674 - 77645263 on Build GRCh38
UniGene ID:
Hs.534699
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS189569
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SCEL
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160706.1 NP_001154178.1 19 1293
NM_003843.3 NP_003834.3 20 1359
NM_144777.2 NP_659001.2 21 1419
XM_005266578.1 XP_005266635.1 20 1330
XM_006719882.1 XP_006719945.1 20 1305
XM_006719884.1 XP_006719947.1 20 1299
XM_006719885.1 XP_006719948.1 19 1245
XM_011535281.1 XP_011533583.1 21 1362
XM_011535282.1 XP_011533584.1 21 1362
XM_011535283.1 XP_011533585.1 21 1365
XM_011535284.1 XP_011533586.1 21 1359
XM_011535285.1 XP_011533587.1 20 1375
XM_011535286.1 XP_011533588.1 20 1305
XM_011535287.1 XP_011533589.1 20 1302
XM_011535288.1 XP_011533590.1 19 1229
XM_011535289.1 XP_011533591.1 19 1242
XM_011535290.1 XP_011533592.1 21 1365
XM_011535291.1 XP_011533593.1 21 1365
XM_017020805.1 XP_016876294.1 21 1365
GenBank mRNA AF045941.1 - 20 1275
AK025320.1 - 21 1333
AK222720.1 - 21 1333
AK301659.1 - 19 1207
BC020726.1 - 17 2131
BC047536.1 - 20 1434 5271857

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

epidermis development
embryo development
keratinocyte differentiation
protein binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header