This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM107B SIRNA Assays ›
Gene Symbol:
FAM107B
Gene Name:
family with sequence similarity 107 member B
Gene Aliases:
C10orf45
Chromosome Location:
Chr.10: 14518557 - 14774897 on Build GRCh38
UniGene ID:
Hs.446315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS188791
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM107B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282695.1
NP_001269624.1
4
927
NM_001282696.1
NP_001269625.1
3
403
NM_001282697.1
NP_001269626.1
2
337
NM_001282698.1
NP_001269627.1
3
651
NM_001282699.1
NP_001269628.1
2
585
NM_001282700.1
NP_001269629.1
2
303
NM_001282701.1
NP_001269630.1
2
424
NM_001282702.1
NP_001269631.1
3
490
NM_001282703.1
NP_001269632.1
2
357
NM_001320735.1
NP_001307664.1
3
369
NM_001320736.1
NP_001307665.1
3
629
NM_001320737.1
NP_001307666.1
3
423
NM_001320738.1
NP_001307667.1
3
389
NM_001320739.1
NP_001307668.1
2
389
NM_001320740.1
NP_001307669.1
2
181
NM_001320741.1
NP_001307670.1
2
282
NM_031453.3
NP_113641.2
3
861
XM_005252616.3
XP_005252673.1
3
258
XM_011519735.1
XP_011518037.1
4
375
XM_017016746.1
XP_016872235.1
3
249
XM_017016747.1
XP_016872236.1
2
226
XM_017016748.1
XP_016872237.1
2
261
XM_017016749.1
XP_016872238.1
2
323
GenBank mRNA
AK074275.1
-
1
120
AK127413.1
-
3
861
AK289959.1
-
3
651
AL136885.1
-
2
347
BC004872.2
-
1
131
BC064407.1
-
2
307
BC072452.1
-
3
403
BC144707.1
-
4
735
BF979432.1
-
2
397
CR533466.1
-
1
102
DA116139.1
-
2
181
DA269906.1
-
2
207
DA305421.1
-
3
300
DA340804.1
-
3
350
DA390740.1
-
2
344
DA409460.1
-
2
363
DA412617.1
-
3
397
DA543225.1
-
2
186
DA813465.1
-
2
210
DA922675.1
-
2
282
DA954468.1
-
2
303
DA962195.1
-
2
211
DB194372.1
-
2
323
DB198416.1
-
3
335
Back To Top
More Information
Gene Ontology Categories:
Function(s)
sensory perception of sound
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0