This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LY6G6C SIRNA Assays ›
Gene Symbol:
LY6G6C
Gene Name:
lymphocyte antigen 6 complex, locus G6C
Gene Aliases:
C6orf24, G6c, NG24
Chromosome Location:
Chr.6: 31718648 - 31721734 on Build GRCh38
UniGene ID:
Hs.732272
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS188477
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LY6G6C
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025261.2 NP_079537.1 3 328
GenBank mRNA AJ315533.1 - 3 273
AJ315534.1 - 4 384
AY359119.1 - 3 328
BC036302.1 - 3 303 4776231

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header