This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBXN8 SIRNA Assays ›
Gene Symbol:
UBXN8
Gene Name:
UBX domain protein 8
Gene Aliases:
D8S2298E, REP8, UBXD6
Chromosome Location:
Chr.8: 30729131 - 30767006 on Build GRCh38
UniGene ID:
Hs.153678
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS188288
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UBXN8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282189.1 NP_001269118.1 8 603
NM_001282199.1 NP_001269128.1 7 591
NM_005671.3 NP_005662.1 8 714
XM_011544655.2 XP_011542957.1 10 1791
XM_017013862.1 XP_016869351.1 9 903
GenBank mRNA AK296600.1 - 8 888
AY302140.1 - 8 578
BC020694.1 - 8 698 4772692
BI755534.1 - 7 567
D83767.1 - 8 704
D83768.1 - 6 441

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

single fertilization
ER-associated ubiquitin-dependent protein catabolic process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0