This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAGIX SIRNA Assays ›
Gene Symbol:
MAGIX
Gene Name:
MAGI family member, X-linked
Gene Aliases:
JM10, PDZX
Chromosome Location:
Chr.X: 49162569 - 49168157 on Build GRCh38
UniGene ID:
Hs.193170
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS188269
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MAGIX
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099681.2 NP_001093151.2 3 258
NM_001099682.2 NP_001093152.2 3 258
NM_024859.3 NP_079135.3 3 258
XM_005278062.4 XP_005278119.1 3 342
XM_005278063.4 XP_005278120.1 3 341
XM_005278064.4 XP_005278121.1 2 118
XM_005278065.3 XP_005278122.1 2 206
XM_006724560.3 XP_006724623.1 3 342
XM_011543985.2 XP_011542287.1 3 339
XM_011543986.2 XP_011542288.1 2 93
XM_011543987.2 XP_011542289.1 2 114
GenBank mRNA AI369342.1 - 3 329
AK025340.1 - 2 40
AK094251.1 - 1 790
AK096058.1 - 2 1280
BC027995.1 - 3 227
DA072017.1 - 2 40

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0