This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF205 SIRNA Assays ›
Gene Symbol:
ZNF205 ZNF205-AS1
Gene Name:
zinc finger protein 205
ZNF205 antisense RNA 1
Gene Aliases:
RhitH, ZNF210, Zfp13
-
Chromosome Location:
Chr.16: 3112562 - 3120517 on Build GRCh38
Chromosome Location:
Chr.16: 3110460 - 3115598 on Build GRCh38
UniGene ID:
Hs.592088
Hs.669490
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID HSS187847
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF205
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042428.1 NP_001035893.1 3 270
NM_001278158.1 NP_001265087.1 3 589
NM_003456.2 NP_003447.2 3 338
XM_005255558.2 XP_005255615.1 3 305
GenBank mRNA AF060865.1 - 3 295
AK313009.1 - 3 327
CB152017.1 - 3 338
Gene Symbol: ZNF205-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024166.1 1 163
NR_024167.1 1 163

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of hydrogen peroxide biosynthetic process
positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm