siRNA Details
| Gene Symbol: LINC01530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034159.1 | 2 | 579 | |||
| GenBank mRNA | AK054965.1 | - | 2 | 576 | ||

LINC01530| Gene Symbol: LINC01530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034159.1 | 2 | 579 | |||
| GenBank mRNA | AK054965.1 | - | 2 | 576 | ||