This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GXYLT2 SIRNA Assays ›
Gene Symbol:
GXYLT2
Gene Name:
glucoside xylosyltransferase 2
Gene Aliases:
GLT8D4
Chromosome Location:
Chr.3: 72848522 - 72975374 on Build GRCh38
UniGene ID:
Hs.710275
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS186527
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GXYLT2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080393.1 NP_001073862.1 4 803
XM_011534067.2 XP_011532369.1 4 669
XM_011534068.1 XP_011532370.1 4 688
XM_017007124.1 XP_016862613.1 4 673
GenBank mRNA BC127733.1 - 3 449 40127813

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

O-glycan processing
UDP-xylosyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0