This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NBPF6 SIRNA Assays ›
Gene Symbol:
NBPF6
Gene Name:
neuroblastoma breakpoint family member 6
Gene Aliases:
-
Chromosome Location:
Chr.1: 108421496 - 108471903 on Build GRCh38
UniGene ID:
Hs.712226
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS185867
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NBPF6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143987.1 NP_001137459.1 16 2194
NM_001143988.1 NP_001137460.1 15 2107
XM_005271144.3 XP_005271201.1 15 2231
XM_011542012.2 XP_011540314.1 20 3560
XM_011542014.2 XP_011540316.1 15 2869
XM_017002148.1 XP_016857637.1 20 2695

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0