This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF574 SIRNA Assays ›
Gene Symbol:
ZNF574
Gene Name:
zinc finger protein 574
Gene Aliases:
FP972
Chromosome Location:
Chr.19: 42068477 - 42081567 on Build GRCh38
UniGene ID:
Hs.13323
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS185534
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF574
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022752.5 NP_073589.4 2 878
XM_005259159.2 XP_005259216.1 1 741
XM_011527228.2 XP_011525530.1 2 1071
XM_011527229.2 XP_011525531.1 2 794
XM_011527230.2 XP_011525532.1 2 746
XM_017027149.1 XP_016882638.1 2 1214
GenBank mRNA AF447880.1 - 2 822
AK027895.1 - 2 781
AK131369.1 - 2 1214
AK131418.1 - 2 1364
BC001184.1 - 2 767 3502564

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0