This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF674 SIRNA Assays ›
Gene Symbol:
ZNF674
Gene Name:
zinc finger protein 674
Gene Aliases:
MRX92, ZNF673B
Chromosome Location:
Chr.X: 46497725 - 46545459 on Build GRCh38
UniGene ID:
Hs.636105
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS184661
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF674
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039891.2 NP_001034980.1 6 1335
NM_001146291.1 NP_001139763.1 6 1317
NM_001190417.1 NP_001177346.1 6 1320
XM_011543941.2 XP_011542243.1 6 1332
XM_011543943.2 XP_011542245.1 6 1334
XM_011543944.2 XP_011542246.1 4 1142
XM_011543945.2 XP_011542247.1 3 1091
XM_017029728.1 XP_016885217.1 7 1487
XM_017029729.1 XP_016885218.1 7 1509
XM_017029730.1 XP_016885219.1 7 1506
XM_017029731.1 XP_016885220.1 3 1091
XM_017029732.1 XP_016885221.1 7 1484
XM_017029733.1 XP_016885222.1 7 1486
GenBank mRNA AK295054.1 - 6 1249
AY971607.1 - 6 1319
BX648127.1 - 1 651

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0