This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OTOR SIRNA Assays ›
Gene Symbol:
OTOR
Gene Name:
otoraplin
Gene Aliases:
FDP, MIAL1
Chromosome Location:
Chr.20: 16748353 - 16752164 on Build GRCh38
UniGene ID:
Hs.41119
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS183603
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OTOR
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020157.3 NP_064542.1 3, 4 414
GenBank mRNA AF233261.1 - 3, 4 409
AF243505.1 - 3, 4 372
AJ242552.1 - 3, 4 365
AY359082.1 - 3, 4 402
BC101688.1 - 3, 4 414 8069194
BC101690.1 - 3, 4 414 8069196

Back To Top

More Information


Gene Ontology Categories:

Function(s)

cartilage condensation
sensory perception of sound

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header