This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other H2AFJ SIRNA Assays ›
Gene Symbol:
H2AFJ
Gene Name:
H2A histone family member J
Gene Aliases:
H2AJ
Chromosome Location:
Chr.12: 14774336 - 14778002 on Build GRCh38
UniGene ID:
Hs.524280
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS183252
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: H2AFJ
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177925.3 NP_808760.1 1 508
NR_027716.2 1 508
GenBank mRNA AK303301.1 - 2 385
AL133626.1 - 1 262
BC003602.1 - 1 429 3354610
BU594879.1 - 1 486
DA808847.1 - 1 509

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing
biological_process
molecular_function
DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0